missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Water Jet Pump
Used for withdrawing liquids and gases, and for creating a vacuum. Buerkle™ Water Jet Pumps offer simple operation and easy cleaning.
Brand: Buerkle™ 9668-0200
74.09 EUR valid until 2025-03-31
Use promo code "24119" to get your promotional price.
This item is not returnable.
View return policy
Description
- Very low water consumption
- Constant pressure and high suction
- Suction tube connection: Olive 6-9mm outer diameter, screw cap GL 14
- Water consumption at 3.5 bar flow pressure and 12°C water temperature: approx. 190L/hour
- Suction capacity at 3.5 bar flow pressure and 12°C water temperature: 400L/hour evacuation time for a 1L pressure vessel from normal pressure to 100 mbar: 60 sec.
- Vacuum connection can be unscrewed and supplied adapters provide a variety of connection options to the water supply
Specifications
Olive 6-9 mm outer diameter, Screw Cap GL 14 | |
190 L/hr | |
Water Jet Pump | |
3.5 bar |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buerkle™ Water Jet Pump > With Reducing piece to 1/2 in.
Spot an opportunity for improvement?Share a Content Correction