Learn More
Aalborg™ Water Direct Glass Flow Tube
Product Code. p-8008842
Click to view available options
Waterflow Rate:
1.2L/min.
100mL/min.
200mL/min.
20mL/min.
250mL/min.
2L/min.
500mL/min.
50mL/min.
Width (Metric):
65 mm
150 mm
180 mm
Unit Size:
Each
Product Code. 11778258
Brand: Aalborg™ FT06406STVN
Only null left
Add to basket
Add to basket
Aalborg™ Water Direct Glass Flow Tube completes a flowmeter, which is used in fluid systems to indicate the rate of flow of the fluid.
  • Direct reading tubes are straightforward.
  • The measurement scale on each of these tubes reads actual flow at standard conditions in a choice of English or Metric units.
  • Obtaining actual flow rates with these tubes requires the use of a reference scale flow correlation table.
  • Reference scale tubes are useful when measuring flow rates for gases other than air and/or for non-standard conditions.
Specifications
Airflow Rate 1.2 L/min.
Waterflow Rate 1.2L/min.
Type Water Direct Flow Tube
Width (Metric) 65 mm