Learn More
DWK Life Sciences Watch Glass Dish, made from soda-lime glass, fused rim
Shop All DWK Life Sciences Products
Click to view available options
Diameter (Metric):
40 mm
50 mm
60 mm
70 mm
80 mm
90 mm
100 mm
120 mm
125 mm
150 mm
200 mm
250 mm
Unit Size:
Each
Pack of 10
Description
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility. Watch glasses, for example, are concave curved. The small trough created in this way allows liquids or solids to be filled in.
- Reduced risk of injury due to fused rim
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Material | Soda Lime Glass |
| For Use With (Application) | Weighing; Covering; Pretreatments; Drying of materials; Available in Boro 3.3 or soda-lime glass |
| No. per Pack | 10/Pk. |
| Diameter (Metric) | 150 mm |