Learn More
Gosselin™ Waste and Sharps Container, White Hinged Cap
Click to view available options
Capacity (Metric):
600 mL
800 mL
1.1 L
1.8 L
4.5 L
7 L
Quantity:
25/Cs.
42/Cs.
45/Cs.
60/Cs.
65/Cs.
70/Cs.
Unit Size:
Pack of 25
Pack of 42
Pack of 45
Pack of 60
Pack of 65
Pack of 70
Description
- Polypropylene construction
- Suitable for waste and sharps
- NFX 30-5111 compliant
- Hinged cap
Specifications
Specifications
| Capacity (Metric) | 4.5 L |
| Certifications/Compliance | NFX 30-5111 |
| Closure Type | Hinged Top |
| Closure Color | White |
| Color | Yellow |
| Quantity | 42/Cs. |