Learn More
Ekastu Safety™ Eye Wash Bottle Wall Cabinet with Transparent Lid
Provides immediate access to Eye Wash Bottles when placed in significant areas. EKASTU Safety™ Eye Wash Bottle Wall Cabinet with Transparent Lid mounts easily and holds one wash bottle.
Brand:  Ekastu Safety™ 277951
Product Code. 10121151
  • € 47.00 / Each
Estimated Shipment: 24-04-2025
to see stock.
Explore more special offers
Description
  • Immediate availability of Eye Wash Bottles on significant places
  • Protect the Eye Wash Bottles for faulting
  • Fast to open transparent lid
  • Very robust and durable
  • Easy wall mounting
  • For one eye wash bottle
Specifications
Specifications
Holds One Eyewash Bottle, Transparent Lid
Wall
Box
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Ekastu Safety™ Eye Wash Bottle Wall Cabinet with Transparent Lid > For Use With 1 Eye wash bottle