missing translation for 'onlineSavingsMsg'
Learn More
VU 152100, Tocris Bioscience™ Product Code.: 18045482

VU 152100, Tocris Bioscience™

Product Code. 18045482
50 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18045482

Brand: Tocris Bioscience 3383/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Positive allosteric modulator of M4 receptors

Selective positive allosteric modulator of M4 muscarinic acetylcholine receptors (mAChRs) (EC50 = 380 nM). Induces 21-fold shift in ACh potency at M4 receptor. Displays no activity at other mAChR subtypes.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 409351-28-6
Molecular Formula C18H19N3O2S
Molecular Weight (g/mol) 341.429
InChI Key MDNWGCQSCGNTKH-UHFFFAOYSA-N
Synonym 3-amino-n-4-methoxyphenyl methyl-4,6-dimethylthieno 2,3-b pyridine-2-carboxamide, 3-amino-n-4-methoxybenzyl-4,6-dimethylthieno 2,3-b pyridine carboxamide, kinome_1269, d0k3hy, vu hplc, 3-amino-4,6-dimethyl-n-p-anisyl-thieno 2,3-b pyridine-2-carboxamide, 3-amino-4,6-dimethylthiopheno 2,3-b pyridin-2-yl-n-4-methoxyphenyl methyl carboxamide, 3-amino-n-4-methoxyphenyl methyl-4,6-dimethyl-2-thieno 2,3-b pyridinecarboxamide, 3-amino-n-4-methoxyphenyl methyl-4,6-dimethyl-thieno 2,3-b pyridine-2-carboxamide, 3-amino-n-4-methoxyphenyl methyl-4,6-dimethylthieno 5,4-b pyridine-2-carboxamide
PubChem CID 864492
IUPAC Name 3-amino-N-[(4-methoxyphenyl)methyl]-4,6-dimethylthieno[2,3-b]pyridine-2-carboxamide
SMILES CC1=CC(=NC2=C1C(=C(S2)C(=O)NCC3=CC=C(C=C3)OC)N)C

Specifications

Quantity 50 mg
Formula Weight 341.43
Percent Purity >98%
Chemical Name or Material VU 152100
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
VU 152100, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.