Learn More
Tocris Bioscience™ VU 0463271
Potent and selective KCC2 inhibitor
Brand: Tocris Bioscience™ 4719/50
Additional Details : CAS : 1391737-01-1 Weight : 0.00970kg
Description
VU 0463271 is a potent and selective inhibitor of the neuronal K-Cl cotransporter, KCC2 (IC50 = 61 nM); displays >100-fold selectivity versus the Na-K-2Cl cotransporter 1 (NKCC1) and no activity against a panel of 68 GPCRs, ion channels and transporters.Specifications
N-Cyclopropyl-N-(4-methyl-2-thiazolyl)-2-[(6-phenyl-3-pyridazinyl)thio]acetamide | |
1391737-01-1 | |
50 mg |