Learn More
Fisherbrand™ Vortex Mixers Tube Holders
Shop All Fisherbrand Products
Click to view available options
No. of Positions:
1
18
24
Unit Size:
Each
Specifications
Specifications
| Holds | 1.5 to 2 mL |
| Product Type | Microtube Holder |
| Dimensions (L x W) | 9 x 9 in. (229 x 229 mm) |
| Length (English) | 9 in. |
| Length (Metric) | 229 mm |
| Width (English) | 9 in. |
| Width (Metric) | 229 mm |
| For Use With (Equipment) | Vortex mixer |
| No. of Positions | 18 |