Learn More
Hirschmann™ Volumetric Pipets with Attached Syringe
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
15 mL
20 mL
25 mL
50 mL
Unit Size:
Each
Description
- One mark
- Amber stain graduation
Specifications
Specifications
| Material | Soda Lime Glass |
| Graduated | Yes |
| Graduation Color | Brown |
| Quantity | Each |
| Tolerance | 0.015 mL |
| Volume (Metric) | 5 mL |
| Type | Volumetric Pipette Syringe |