Learn More
Hirschmann™ Volumetric Pipets with Attached Syringe
Provides an extremely high degree of volumetric accuracy for your pipetting needs. Hirschmann™ Volumetric Pipets with Attached Syringe come with amber graduation.
Brand: Hirschmann™ 1410133
This item is not returnable.
View return policy
Description
- One mark
- Amber stain graduation
Specifications
Soda Lime Glass | |
Brown | |
0.008 mL | |
Volumetric Pipette Syringe |
Yes | |
Each | |
1 mL |
Product Content Correction
Product Title
Hirschmann™ Volumetric Pipets with Attached Syringe > Capacity: 1 mL