Learn More
Hirschmann™ Volumetric Measuring Device Fitting
Product Code. p-7162699
Click to view available options
For Use With (Equipment):
Schilling Fast-Flow Burettes, 15 - 50 mL
Schilling Fast-Flow Burettes, 5 - 10 mL
Unit Size:
Each
Product Code. 10069770
Brand: Hirschmann™ 8900710
Only null left
Add to basket
Add to basket
Fitting for Dr. Schilling fast-flow burettes
  • For Schilling burettes
Specifications
For Use With (Equipment) Schilling Fast-Flow Burettes, 5 - 10 mL
No. per Pack 1 Each
Type Buret Fitting