missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 2 Marks and Blue Graduations
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 2 Marks and Blue Graduations allow accurate measurements of a volume of solution.
Brand: Poulten & Graf GmbH 1 2323602
This item is not returnable.
View return policy
Description
- With 2 marks
- Blue graduations
- Conformity approved
- With batch certificate
Specifications
EN ISO 648 | |
Soda Lime Glass | |
Yes | |
Ring marks | |
0.02 mL | |
Two-mark Bulb Pipette |
AS | |
Non-sterile | |
Blue | |
5 | |
10 mL | |
Reusable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction