Learn More
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 2 Marks and Blue Graduations
Product Code. 10476691
Click to view available options
Volume (Metric):
3 mL
4 mL
5 mL
10 mL
15 mL
25 mL
50 mL
100 mL
Unit Size:
Each
Pack of 5
Product Code. 10476691
Brand: Poulten & Graf GmbH 1 2323002
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 2 Marks and Blue Graduations allow accurate measurements of a volume of solution.
  • With 2 marks
  • Blue graduations
  • Conformity approved
  • With batch certificate
Specifications
Certifications/Compliance EN ISO 648
Class AS
Material Soda Lime Glass
Sterility Non-sterile
Graduated Yes
Graduation Color Blue
Graduation Type Ring marks
Quantity 5
Tolerance 0.015 mL
Volume (Metric) 4 mL
Type Two-mark Bulb Pipette
Disposable Reusable