Learn More
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 2 Marks and Amber Graduations
Product Code. p-7116431
Click to view available options
Volume (Metric):
1 mL
3 mL
4 mL
5 mL
10 mL
15 mL
20 mL
50 mL
100 mL
Unit Size:
Each
Pack of 5
Product Code. 15952130
Brand: Poulten & Graf GmbH 1 2324504
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 2 Marks and Amber Graduations allow accurate measurements of a volume of solution.
  • With 2 marks
  • Amber graduations
  • Conformity approved
  • With batch certificate
Specifications
Certifications/Compliance EN ISO 648
Class AS
Material Soda Lime Glass
Sterility Non-sterile
Graduated Yes
Graduation Color Amber
Graduation Type Ring marks
Quantity 5
Tolerance 0.05 mL
Volume (Metric) 50 mL
Type Two-mark Bulb Pipette
Disposable Reusable