Learn More
Fisherbrand™ Viton Rubber Tubing
Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Inner:
1 mm
2 mm
3 mm
4 mm
5 mm
6 mm
8 mm
10 mm
12 mm
Thickness (Metric):
1 mm
1.5 mm
2 mm
2.5 mm
3 mm
Unit Size:
5m
Specifications
Specifications
| Diameter (Metric) Inner | 6 mm |
| Material | Viton |
| Overall Length (Metric) | 5 m |
| Thickness (Metric) | 1.5 mm |
| For Use With (Application) | Transferring corrosive liquids, even at high temperatures |