Learn More
VistaLab™ Technologies Cap Opener
An easy-to-use, ergonomic solution for opening PCR or microtubes while pipetting
Brand:  VistaLab™ Technologies 9057-4004
Product Code. 11975962
  • € 74.00 / Pack of 3
Explore more special offers
Specifications
Tubes with 8 mm Caps
1 Ea.
3 in.
Ovation electronic single channel and adjustable and fixed volume models
Ideal easy-to-use ergonomic solution for opening PCR or micro tubes while pipetting
3 in.
Manual
Cap Opener Kit
11 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
VistaLab™ Technologies Cap Opener > Cap opener kit