Learn More
Invitrogen™ ViewRNA™ Cell Plus Assay Kit
The ViewRNA Cell Plus Assay is a novel assay that combines immunocytochemistry with ViewRNA technology, a proprietary fluorescent in situ hybridization and sequential branched-DNA amplification technique, to visualize both RNA with single-molecule sensitivity and protein in individual cells.
Brand: Invitrogen™ 88-19000-99
Description
This assay enables simultaneous detection of up to three RNA targets in combination with immunophenotyping for cell surface or intracellular proteins using both indirect and direct immunocytochemistry to allow for detailed characterization of specific cell subpopulations. For simultaneous detection of a fourth RNA target, the ViewRNA ISH Cell 740 Module (Cat. No. QVC0200) can be combined with this kit. It allows analysis of an additional target in the 740 channel (AlexaFluor 750).
This ViewRNA Cell Plus Assay kit contains all the reagents needed to conduct the assay. Target Probe sets for genes of interest and antibodies are sold separately.
Reactivity/species
Probes can be designed for any species. These are the species most frequently used/tested: Armenian hamster, baboon, bovine, canine, cat, chicken, chimpanzee, cow, cynomolgus monkey, donkey, goat, golden Syrian hamster, guinea pig, hamster, horse, human, macaque, monkey, mouse, non-human primate, olive baboon, pig, pigtailed macaque, rabbit, rat, rhesus monkey, sheep
Reported application
Microscopy, immunocytochemistry
Specifications
Primer-probe Detection | |
Microscopy, Immunocytochemistry | |
1 kit |
Kit | |
RNA In Situ Hybridization Assay Kit |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
415951-204 | Certificate of Analysis | 10/04/2025 | |
415951-305 | Certificate of Analysis | 10/04/2025 | |
415951-304 | Certificate of Analysis | 10/04/2025 | |
415951-104 | Certificate of Analysis | 10/04/2025 | |
415951-105 | Certificate of Analysis | 10/04/2025 |