Learn More
OHAUS™ Vessel Holder for Microplate Vortex Mixer
Rubber holder secures Erlenmeyer flasks. Vessel holder also includes a grip mat. Requires Universal Holder.
Brand: OHAUS™ 30400218
44.56 EUR valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
- For use with Ohaus Microplate Vortex Mixer
Specifications
125 and 250 mL Erlenmeyer Flasks | |
Plasitc | |
Vortex Mixer | |
1 Ea. |
Small Vessel Holder | |
Small Vessel Holder 125 & 250 mL Flasks | |
Grip mat |