Learn More
Scientific Industries SI™ Vertical Tube Holder
Click to view available options
Tube Capacity:
15 mL
50 mL
50 mL, 15 mL, microtubes
No. of Positions:
12
3
6
6 x 50 mL, 6 x 15 mL Tubes, 12 Microtubes
Unit Size:
Each
Description
- Holds tubes vertically
- Snaps on to any Vortex-Genie 2 Mixer and the Vortex-Genie Pulse
- Optimized for high speed vortexing a small number of tubes
- No additional attachment necessary
Specifications
Specifications
| Tube Capacity | 50 mL |
| For Use With (Equipment) | Vortex-Genie 2, Vortex-Genie Pulse |
| No. of Positions | 3 |