Learn More
Scientific Industries SI™ Vertical Microtube Holder
Product Code. p-6966051
Click to view available options
No. of Positions:
9
24
Unit Size:
Each
Product Code. 15587365
Brand: Scientific Industries SI™ 83041243
Only null left
Add to basket
Add to basket
Requires no additional attachment for use. Scientific Industries SI™ Vertical Microtube Holder snaps microtubes into place.
  • Holds tubes vertically
  • Snaps on to any Vortex-Genie 2 Mixer and the Vortex-Genie Pulse
  • Optimized for high speed vortexing a small number of tubes
  • No additional attachment necessary
Specifications
For Use With (Equipment) Vortex-Genie 2 Mixer and Vortex-Genie Pulse
No. of Positions 9