Learn More
Kinematica HMC Europe Vertical Autoclaves: HG Series
Self contained top loading autoclaves offer large capacity in a compact unit and easy to use controls.
Brand: Kinematica HMC Europe 21001
This item is not returnable.
View return policy
Description
Programmable top-loading autoclaves/sterilizers
- Use steam sterilization
- Six sizes from 25 to 110 liters
- Low height, easy-to-load
- Portable — require no plumbing to operate but can be plumbed
- Complete line of baskets, inserts and cans available separately
- HG Series (advanced features)
- Many upgrades available depending on your needs
Certifications
CE marked
Services
Two years on parts; One year on labor
Specifications
CE | |
69.1 cm | |
50 L | |
36.4 cm | |
35 in. | |
HG 50 | |
105°C to 135°C | |
62 kg | |
18.97 in. | |
3.8 w | |
45.5 cm |
27.2 in. | |
13.2 gal. | |
14.33 in. | |
Steam sterilization | |
89 cm | |
221°F to 275°F | |
136.68 lb. | |
48.2 cm | |
2.5 psi | |
17.91 in. |
Product Content Correction
Product Title
Kinematica HMC Europe Vertical Autoclaves: HG Series > HG Series; 50L