Learn More
Thermo Scientific™ Versette™ Automated Liquid Handler
Product Code. 15443770 Shop All Thermo Scientific Products
Click to view available options
Description:
6-position stage for Versette, guarding included
96- and 384- channel pipetting module for use with 96- and 384-channel Versette pipetting heads
Versette Base Unit Only
Versette instrument (includes base, pipetting module and a 6-position stage)
Versette system for MSIA application (includes base, pipetting module, 6-position stage, 96-channel pipetting head for volumes of 5-300μl and a serial dilute magazine 96/300μl)
Unit Size:
Each
Product Code. 15443770
Brand: Thermo Scientific™ 650INSTR
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Support a wide range of applications with this compact handler, featuring user-friendly programming, reliable performance and a choice of 96- or 384-channel pipetting heads.
Support a wide range of applications with the Thermo Scientific™ Versette™ Automated Liquid Handler, a compact handler featuring user-friendly programming, reliable performance and a choice of 96- or 384-channel pipetting heads. An ideal choice for the liquid handling needs of most laboratories, it supports 96/384 plate replication, plate stamping and serial dilution procedures.
Support a wide range of applications, including high-throughput Mass Spectrometric Immunoassay (MSIA), plate stamping, plate reformatting, serial dilution and many other applications.
96-and 384-channel pipetting heads:
  • Total volume range 0.5-300μL
  • All pipetting heads include RFID tags for self-identification
Consumables for optimized performance:
  • All pipetting heads utilize Thermo Scientific™ D.A.R.T.'S
Compact size:
  • Six position stage uses a unique dual-level design to minimize space requirements
  • Fits on standard bench
User-friendly programming options:
  • Simple pipetting procedures can be created using visual on-board graphical display
  • For more complex protocols and advanced editing, use Thermo Scientific™ ControlMate™ software included with purchase of the unit
  • Intuitive software; after minimal training, no specialist needed for protocol writing and revisions
  • Free WebEx ControlMate SW tutorial and applications support
TRUSTED_SUSTAINABILITY
Specifications
Product Type Versette Instrument
Description Versette instrument (includes base, pipetting module and a 6-position stage)
Channels 96, 384
Volume (Metric) 0.5 to 300 μL
For Use With (Equipment) 96-, 384- well plates with different heights, storage tubes in rack (Thermo Scientific 0.5 to 1.4mL)
Interface RS-232, USB
Operating System Windows XP, Windows Vista (32-and 64-bit), Windows 7 (32- and 64-bit), WIN 10
Width (English) 27 in.
Width (Metric) 68 cm
Height (English) 27 in.
Height (Metric) 68 cm
Depth (English) 22 in.
Depth (Metric) 55 cm
Weight (English) 120 lb. (base system - heads/pipetting module not included), 80 lb. (base - tower), 40 lb. (6 position stage), 26 lb. (NTC pipetting module)
Weight (Metric) 54.4 kg (base system - heads/pipetting module not included), 36 kg (base - tower), 18.1 kg (6 position stage), 11.8 kg (NTC pipetting module)
Electrical Requirements 100 to 240V 50/60Hz
Voltage 100/240 V
Frequency 50/60 Hz