Learn More
3M™ Versaflo™ M-Series Faceshields and Helmets
Product Code. p-200087361
Click to view available options
Includes:
Comfort Faceseal
Flame Resistant Faceseal
Flame Resistant Poly Faceseal
Flame Resistant Shroud
Highly Durable Shroud
Unit Size:
Each
Model:
M-406
Product Code. 17825666
Brand: 3M™ 7100258667
Only null left
Add to basket
Add to basket
3M™ Versaflo™ M-Series Faceshields and Helmets can help protect your workers from a variety of respiratory and impact hazards while maintaining comfort. They are are lightweight, compact and easy to maintain.
  • M-Series are specifically designed to work together with your 3M™ Versaflo™ Powered Air Respirator and 3M™ Supplied Air Respirator Systems.
  • Deflector allows users to direct the airflow inside the headtop to increase comfort.
  • Fully-adjustable suspension for customized fit and weight distribution.
  • Accessory options availalble for protection to help handle the environments you encounter.
  • Visor combines good peripheral and downward vision.
  • Specific headtops can be used with a range of 3M™ Peltor™ hearing protection as well as an attachable microphone for ease of communication.
TRUSTED_SUSTAINABILITY
Specifications
Color Light Gray
Design Type Comfort Faceseal
Disposable Reusable
Filter Type None
Material Polyethylene
Product Type Hood with Faceseal
Size Standard
With Exhalation Valve No
Includes Comfort Faceseal