Learn More
3M™ Versaflo™ M-Series Faceshields and Helmets
Click to view available options
Includes:
Comfort Faceseal
Flame Resistant Faceseal
Flame Resistant Poly Faceseal
Flame Resistant Shroud
Highly Durable Shroud
Unit Size:
Each
Model:
M-406
Description
- M-Series are specifically designed to work together with your 3M™ Versaflo™ Powered Air Respirator and 3M™ Supplied Air Respirator Systems.
- Deflector allows users to direct the airflow inside the headtop to increase comfort.
- Fully-adjustable suspension for customized fit and weight distribution.
- Accessory options availalble for protection to help handle the environments you encounter.
- Visor combines good peripheral and downward vision.
- Specific headtops can be used with a range of 3M™ Peltor™ hearing protection as well as an attachable microphone for ease of communication.
Specifications
Specifications
| Color | Black |
| Design Type | Flame Resistant |
| Disposable | Reusable |
| Filter Type | None |
| Material | Flame Resistant Polyester |
| Product Type | Respirator Helmet |
| Size | Standard |
| With Exhalation Valve | Yes |
| Includes | Flame Resistant Faceseal |
| Visor Type | Polycarbonate Visor |