Learn More
VD3-d6, MedChemExpress
MedChemExpress VD3-d6 (Vitamin D3-26,26,26,27,27,27-d6) is the deuterium labeled Vitamin D3; tools for determination of Vitamin D3 metabolites in human serum.
Brand: Medchem Express HY-15331
This item is not returnable.
View return policy
Description
VD3-d6 (Vitamin D3-26,26,26,27,27,27-d6) is the deuterium labeled Vitamin D3Specifications
VD3-d6 | |
1 mg | |
Research | |
Vitamin D3-26,26,26,27,27,27-d6 | |
O[C@@H](CCC/1=C)CC1=C\C=C2[C@@](CC[C@@H]3[C@@H](CCCC(C([2H])([2H])[2H])C([2H])([2H])[2H])C)([H])[C@]3(C)CCC\2 | |
390.67 | |
Research | |
Solid |
118584-54-6 | |
Light Yellow | |
C27H38D6O | |
DMSO : 100 mg/mL (255.97 mM; Need ultrasonic) | |
390.67 | |
98.0% | |
Neuroscience-Neuromodulation |
Research purposes only