Learn More
VD2-d3, MedChemExpress
MedChemExpress VD2-d3 is the deuterium labeled vitamin D2.
Brand: Medchem Express HY-15330
This item is not returnable.
View return policy
Description
VD2-d3 is the deuterium labeled vitamin D2.Specifications
VD2-d3 | |
5 mg | |
Research | |
DMSO : 20 mg/mL (50.04 mM; ultrasonic and warming and heat to 80°C) | |
399.67 | |
95.46% | |
Metabolism-sugar/lipid metabolism |
1217448-46-8 | |
White | |
C28H41D3O | |
O[C@@H](CCC/1=C([2H])\[2H])CC1=C([2H])\C=C2[C@@](CC[C@@H]3[C@@H](/C=C/[C@H](C)C(C)C)C)([H])[C@]3(C)CCC\2 | |
399.67 | |
Research | |
Solid |
Research purposes only