Learn More
MedChemExpress VCH-916
MedChemExpress VCH-916 is a novel nonnucleoside HCV NS5B polymerase inhibitor.
Brand: MedChemExpress HY-13465
This item is not returnable.
View return policy
Specifications
497.73 g/mol | |
4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture) | |
98.48% | |
DMSO : ≥ 39 mg/mL (78.36 mM) | |
1200133-34-1 |
Reference compound | |
Room temperature | |
100 mg | |
COVID-19-anti-virus |