Learn More
KNF™ Diaphragm Vacuum Pump System SC 920 G
Speed-controlled diaphragm vacuum pump system for safe operation.
Brand:  KNF™ 306473/117056
Product Code. 15810264
  • € 7100.00 / Each
Estimated Shipment: 23-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Automatic, accurate recognition and monitoring of the boiling point using the integrated ramp function.
  • Speed-controlled.
  • Integrated gas ballast valve.
Specifications
Specifications
2 Mbar abs.
Pump head: PPS, Diaphragm: PTFE-coated, Valves: FFPM
40°C
366 x 423 x 294 mm
15.2 kg
1.26 m³/hr (at atm. pressure 1.26)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
KNF™ Diaphragm Vacuum Pump System SC 920 G > Flow rate: 1.26 m3/h at atm. pressure 1.26