Learn More
Welch Ilmvac™ Vacuum Pump Oil
Product Code. p-8012726 Shop All Welch Ilmvac Products
Click to view available options
Volume (Metric):
5 L
20 L
Unit Size:
20 L
5 L
Product Code. 10420853
Brand: Welch Ilmvac™ 800124
One and two-stage rotary vane pump. Fisherbrand™ Vacuum Pump Oil designed for use with the range of Fisherbrand two-stage rotary vane pumps and with most other one and two-stage rotary vane pumps.
  • Generic mineral oil
  • Use in most one and two stage rotary vane pumps
  • 20L drum
  • Density: 0.888g/mL
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vacuum Pump Oil
Pressure 10 - 5 mbar
Viscosity 118 cSt viscosity at 40°C
Volume (Metric) 20 L