Learn More
Greiner Bio-One Vacuette Tube™, Lavender Cap-White Ring, K2E K2EDTA, Sterile
Inner wall is coated with anticoagulant that binds calcium ions and interrupts clotting.
Brand:  Greiner Bio-One 454047
Product Code. 15402530
  • € 75.25 / Pack of 50
Estimated Shipment: 20-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Inner wall is coated with spray-dried K2E K2EDTA as an anticoagulant to bind calcium ions and interrupt the clotting cascade.
  • Inner wall is coated with spray-dried K2E K2EDTA as an anticoagulant to bind calcium ions and interrupt the clotting cascade.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vacuette Tube
Sterile
Lavender with White Ring
13 mm
K2 E K2 EDTA
2 mL
75 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers