Learn More
Buerkle™ V2A Stainless Steel Scoop
Use this Buerkle™ V2A Stainless Steel Scoop as a measuring scoop in the laboratory. It is also suitable for use with foodstuffs
Brand: Buerkle™ 5370-1000
This item is not returnable.
View return policy
Description
Stainless steel V2A scoop for sampling.- Stainless steel V2A (1.4301)
- Sterilizable
- Robust
Specifications
Measuring Scoop | |
Autoclavable | |
Non-sterile | |
Suitable for Use with Foodstuffs |
Stainless Steel (V2A) | |
1000 mL | |
AISI 304, V2A (1.4301) |
Product Content Correction
Product Title
Buerkle™ V2A Stainless Steel Scoop > Capacity: 1000mL; Diameter: 100mm