Learn More
UVP UVP XX-15 Exposure Stand
Click to view available options
For Use With (Equipment):
Model XX-15 bench lamp
Unit Size:
Each
Description
- XX-15 Lamp models can be attached to the Exposure Stand for exposure of materials to ultraviolet
- Shelf is adjustable to different levels to set distances from UV source
Specifications
Specifications
| Depth (English) | 6 in. |
| Depth (Metric) | 152.4 mm |
| For Use With (Equipment) | Model XX-15 bench lamp |
| Height (English) | 13 in. |
| Height (Metric) | 330.2 mm |
| Packaging Quantity | 1 Ea. |
| Width (English) | 20 in. |
| Width (Metric) | 508 mm |
| Product Type | Exposure Stand |