Learn More
UVP White Light Replacement Tubes
Click to view available options
For Use With (Equipment):
14 Type EL series UV lamp
16 Type EL series UV Lamp, High Performance Transilluminator
Model MRL-58 Multiple-ray Lamp
Model TW-43 White Light Transilluminator
Models TLW-20, TMW-20, TW-26 Transilluminator
Wattage:
4 W
6 W
8 W
14 W
Unit Size:
Each
Specifications
Specifications
| Light Source | White light |
| Wattage | 6 W |
| For Use With (Equipment) | 16 Type EL series UV Lamp, High Performance Transilluminator |
| Includes | 6 W cool white tube |
| Length (English) | 8.3 in. |
| Length (Metric) | 21.08 cm |
| Packaging Quantity | 1 Ea. |
| Product Type | Replacement Tube |