Specifications
Lens Coating | Anti-fog |
Lens Tint | Clear |
Product Type | Anti-fog Goggles |
Frame Color | Gray |
No. per Pack | 1 Ea. |
Material | Polycarbonate |
Disposable | Reusable |
Lens Material | Polycarbonate |
Frame Material | Plastic |
UV Protection | Yes |
Show More |
Product Content Correction
Product Title
uvex 9405 Anti-fog Goggle >