Learn More
Uvex™ 1 Safety Shoe S2
Shop All Uvex Products
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
Each
Description
Specifications
Specifications
| Certifications/Compliance | ESD certified, EN ISO 20345: 2011 S2 SRC |
| Color | Black |
| Design | Loafer |
| Material | Microvelour |
| Size | 41 |
| Toe Material | Composite |
| Width (Metric) | 11 mm |
| Gender | Unisex |
| Ankle Style | Trainers |
| Waterproof | No |
| Show More |