Learn More
Cytiva UV Cell
Shop All Cytiva Products
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
U9-L
missing translation for 'unitSize'
Each
Description
A 2mm flow cell for UV-LED monitor U9-L. Included with ÅKTA pure equipped with U9-L. Can be used as second UV monitor.
Specifications
Specifications
| For Use With (Equipment) | U9-L |
| Type | UV Cell |
| Includes | ÄKTA pure equipped with U9-L. Can be used as second UV monitor |
| Quantity | 1 Each |