Learn More
Hirschmann™ USP Calibration Certificate
Click to view available options
Unit Size:
Each
Description
- Individual USP calibration certificate
- Markings with instrument ID
- The quality certificate and the instrument are equipped with an individual serial number
- The certificate provides the following information: the nominal volume, the prescribed tolerance according USP, the actually determined volume of the instrument and the uncertainty of measurement
Specifications
Specifications
| For Use With (Equipment) | Volumetric Glassware, Class A/AS, Confomity Certified acc. to USP |
| Product Type | USP Certificate of Performance |