Learn More
Fisherbrand™ Upright ULT Freezer Racks for Microtiter Plates
Fisherbrand inventory control racks for 96-Well and 384-Well microtiter plates. They fit all standard types of upright ULT freezers.
Brand: Fisherbrand™ UFMP-509L
Description
- Holds up to 96 plates with lid or 120 plates without lid
- Made of high quality stainless steel
- Side opening with handles on the front for easy plate retrieval.
Specifications
Side Access Freezer Rack | |
Stainless Steel | |
Upright ULT Freezer |
Plates with Lid (70) | |
96/384-Well Microtiter Plates | |
466 x 233 x140mm |