Learn More
Liebherr™ Upright Freezer with Shelves
Product Code. 17888306
Click to view available options
Capacity (Metric):
316 L
499 L
573 L
Unit Size:
Each
Product Code. 17888306
Brand: Liebherr™ FFFSG6501
Secure, long-lasting and ultra energy-efficient freezer storage for non temperature-sensitive products, the Liebherr™ Professional Freezer is designed for general purpose applications with Liebherr build-quality you can rely on.
  • Professional upright freezer for general purpose storage, reach-in, 1-section
  • Temperature range -9 / -26°C, Class L1
  • Climate class (energy directive): 7
  • SmartFrost static cooling system (manual defrost): White exterior, commercial-grade moulded polystyrol interior
  • Refrigerant R 290, connection rating 2.0 A, 220-240V/50/1-ph
  • Multifunctional digital controller with high resolution display and connectivity options
  • Self-closing door at <90° and mechanical lock, aluminium door handle
Specifications
Type Professional Freezer
Capacity (Metric) 573 L
Temperature Range -10°C to -26°C
Temperature Sensor NTC
Temperature (Setpoint) -20°C
Door Style Reversible, Self-closing, Fitted Lock
No. of Doors 1
Cabinet Material Scratch-resistant Painted Steel
Chamber Material Stainless Steel
Interior Commercial-grade Molded Polystyrol
Exterior Finish White Powder-coated
Shelves 5
Certifications/Compliance ISO 9001, ISO 14001, CE
Style Upright
Display Touch Control, Digital Display in Door
Casters Optional (80 to 100 mm dia.)
Color White
Condenser Type Low-maintenance Rear-wall Wire-on-tube
For Use With (Application) Storage of Non-sensitive Products
Ports No
Warranty 2 yr. Parts and Labor
Defrost Manual
Model FFFsg 6501
Monitoring Options SmartCoolingHub
Amperage 2.0 A
Plug Type CEE 7/7
Voltage 220/240 V
Phase 1, Single Phase
Wattage 150 W
Energy Usage 1170 kWh/a
Compressor CFC-free
Refrigerant R 290