Learn More
Grant Instruments™ Universal Tray for Grant Shaking Water Baths
Click to view available options
For Use With (Equipment):
LSB12
LSB18
OLS26
Unit Size:
Each
Description
Holds flasks, bottles, and other vessels in place when used with Grant Shaking Water Baths. Includes adjustable springs.
- Universal tray with adjustable springs
- Highly versatile for a variety of vessel types
Specifications
Specifications
| For Use With (Equipment) | LSB18 |
| Type | Universal Tray |