Learn More
Buerkle™ Universal thread adapter
Shop All Bürkle Products
Click to view available options
For Use With:
Buerkle Pumps
Unit Size:
Each
Description
- With G2 connector thread for Bürkle drum screw connections and pumps with immersion tube diameter up to 32 mm
- Material stainless steel AISI 304 and ePTFE seal
- Min. height of the container opening 6 mm, max. 55 mm)
- Scope of delivery: Universal adapter incl. ePTFE seal, mounting key
Specifications
Specifications
| For Use With | Buerkle Pumps |
| Material | Stainless Steel |
| Sterility | Non-sterile |