Learn More
Vitlab™ Universal Model Natural Rubber Pipet Filler
Product Code. 10499382
Click to view available options
Type:
Pipette Filler
For Use With (Equipment):
For pipettes up to 100mL
Material:
Natural Rubber
Unit Size:
Each
Product Code. 10499382
Brand: Vitlab™ 104199
Only null left
Add to basket
Add to basket
Manufactured from natural rubber. Vitlab™ Universal Model Natural Rubber Pipet Filler is used for pipetting with volumetric or measuring pipettes.
  • For pipetting with volumetric or measuring pipettes
  • With 3 valves:
    • Valve A: Air release
    • Valve B: Liquid filling
    • Valve E: Liquid dispensing
Specifications
Material Natural Rubber
Quantity 1 Each
Type Pipette Filler
For Use With (Equipment) For pipettes up to 100mL