Learn More
Abeba™ UNI6 6761 Shoes
Special lining with silver ions absorbs resperation allowing the foot to breathe. Sanitized™ treated.
Brand: Abeba™ 6761/39
This item is not returnable.
View return policy
Description
- CE EN ISO 20347:2012 O2 FO SRC
- Functional leather with honeycomb pattern
- Black
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable Soft Comfort insole M (article no. 350112)
- Sanitized treated
- Antistatic
- Breathable lining with Silver ions
- Instep area with lacing
- Scratch-proof
- Water-repellent
Specifications
CE EN ISO 20347:2012 O2 FO SRC | |
Leather | |
Slip-resistant | |
6761 |
Black | |
39 | |
Polyurethane |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Abeba™ UNI6 6761 Shoes > Size: 39