Learn More
OHAUS™ Uni Block
Shop All Ohaus Products
Click to view available options
For Use With (Equipment):
12 mm Test Tubes
12 mm Vials
15 mm Vials
16 mm Test Tubes
17 mm Vials
20 mm Test Tubes
21 mm Vials
25 mm Test Tubes
28 mm Vials
Unit Size:
Each
Description
Holds test tubes or vials
Specifications
Specifications
| For Use With (Equipment) | 12 mm Vials |
| Type | Uni Block |
| Material | Metal |