Learn More
BICASA Undercounter Drawer Cabinet on Casters
Shop All BICASA Products
Click to view available options
Color:
Anthracite
Blue
Light Gray
Red
Dimensions (D x W x H):
600 x (500 + 20) x 630 mm
600 x (500 + 20) x 760 mm
900 x (500 + 20) x 760 mm
Unit Size:
Each
Description
- Undercounter cabinets on casters
- Metal frame base with 4 swivel casters (Polyamide case and thermoplastic rubber treads), 2 with lockable with brakes - Height mm 100
- Central locking system as standard
- Anti-tilt safety system to open only one drawer at a time
- Body width mm 500 + mm 20 of ergonomic anodized and painted aluminum handle
Specifications
Specifications
| Product Type | Mobile Storage Cabinet |
| Color | Red |
| Cabinet Type | Underbench cabinet |
| Dimensions (D x W x H) | 600 x (500 + 20) x 760 mm |
| No. of Doors | 1 |
| No. of Drawers | 4 |
| Width (English) | 23.6 in. |
| Width (Metric) | 600 mm |
| With Casters | Yes |
| Material | Melamine |