Learn More
Sonics Materials Ultrasonic Probe
Click to view available options
Material:
Titanium Alloy
Unit Size:
Each
Description
- Microtips and probes intensify and radiate the ultrasonic energy into the sample
- Smaller diameter tips produce greater intensity of cavitation, but the energy released is restricted to a narrower, more concentrated field
- Conversely, larger diameter tips produce lower intensity, but the energy is released over a greater area
- The larger the tip diameter, the larger the volume that can be processed, but at reduced intensity
Specifications
Specifications
| Volume Range (Metric) | 250 μL to 10 mL |
| Diameter (Metric) | 3 mm |
| Length (Metric) | 138 mm |
| Autoclavable | Autoclavable |
| Material | Titanium Alloy |