Learn More
Invitrogen™ UltraComp eBeads™ Plus Compensation Beads
Product Code. p-7184690
Click to view available options
No. of Tests:
25
100
Unit Size:
100 tests
2g
Each
Product Code. 16846071
Brand: Invitrogen™ 01333341
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
UltraComp eBeads Plus Compensation Beads are the second generation of UltraComp beads for use in flow cytometry.
  • Binds the spherical particles for use as a single-color compensation controls.
  • Binds with mouse, rat and hamster antibodies, but also bind with rabbit and recombinant human antibodies.
  • Designed for use in compensation with all fluorochromes excited by ultraviolet (355 nm) violet (405 nm), blue (488 nm), green (532 nm), yellow-green (561 nm), and red (633-640 nm) lasers.
  • Compensate in a manner similar to cells when used with Brilliant Violet 785/786-, Brilliant Violet 711-, Super Bright 780- and Super Bright 702-conjugated antibodies.
  • Multispecies compatibility—bind with mouse, rat, hamster, rabbit and recombinant human antibodies
  • Improved fluorophore compatibility
  • Each drop of beads contains two populations: a positive population that bind with any mouse, rat, hamster, rabbit or recombinant human antibody and a negative population that will not bind with antibody.
  • When a fluorochrome-conjugated antibody is added to the beads, both positive and negative populations are observed.
  • This bimodal distribution can be used for single-color compensation controls to accurately setup instrumentation for multicolor flow cytometry experiments.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Flow Cytometry
No. of Tests 25
Product Line eBeads
Quantity 25 tests
Product Type Compensation Beads
RUO use only.