Learn More
Wheaton™ Wheaton™ Ultra Pure OmniFlex Plus Stoppers
Shop All Wheaton Science Products Products
Click to view available options
Diameter (Metric):
13 mm
20 mm
Unit Size:
Pack of 1000
Description
- Autoclavable
- Completely coated with a fluorinated polymer
- Complies with US, European and Japanese Pharmacopeias
Specifications
Specifications
| Diameter (Metric) Inner | 7 mm |
| Diameter (Metric) | 13 mm |
| Autoclavable | Autoclavable |
| Color | Gray |
| Material | Bromobutyl Rubber |
| Product Type | Straight Plug Stopper |