Learn More
SI Analytics™ Ubbelohde Viscometer Not Calibrated
Shop All SI Analytics Products
Click to view available options
Viscosity Range:
0.5 to 3 cSt
10 to 100 cSt
Approximate Constant:
0.003K
0.1K
Size:
0.47mm Diameter
1.13mm Diameter
Unit Size:
Each
Description
- Suitable for transparent liquids manual and automatic measurement, foaming liquids, liquid mixture with highly volatile components, and high or low temperature measurements
- The recommended minimum flow through time is 200 seconds
- Not calibrated
- Without constant for determination of relative viscosity
- Filling quantity of 15 to 20mL
Specifications
Specifications
| Product Type | Viscometer |
| Viscosity Range | 0.5 to 3 cSt |
| Calibrated | Yes |
| Approximate Constant | 0.003K |
| Certifications/Compliance | DIN 51 562 Part 1, ISO/DIS 3105 (BS-IP-SL) |
| Includes | Ring Marks |
| For Use With (Application) | For Determination of Absolute and Relative Kinematic Viscosity of Liquids With Newtonian Flow Behavior |
| Size | 0.47mm Diameter |
| Volume (Metric) | 15 to 20 mL |