Learn More
Poulten Selfe™ U-Tube Reverse Flow Viscometers
Click to view available options
Size:
3
4
Unit Size:
Each
Description
Type BS/IP/RF, IP71, IP319, ASTM D2170.Suitable for transparent and opaque liquids. Nominal overall length 275mm. Approximate sample size 12mL to 25mL. With UKAS certificate of calibration.
Specifications
Specifications
| Product Type | U-Tube Reverse Flow Viscometers |
| Viscosity Range | 6 to 30 cSt |
| ASTM Numbers | D446 |
| Certifications/Compliance | UKAS |
| Size | 3 |
| Volume (Metric) | 12 to 25 mL |